R&D Systems
Manufacturer Code:AF1560SP
Catalog # AF1560SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human PDGF-C Gly230-Gly345 Accession # Q9NRA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Fallotein; FALLOTEIN PDGFC PDGF-C platelet derived growth factor C platelet-derived growth factor C SCDGFSpinal cord-derived growth factor secretory growth factor-like protein VEGF-E; PDGF-C; PDGFC latent form; PDGFC receptor-binding form; Platelet-derived growth factor C; Platelet-derived growth factor C, latent form; Platelet-derived growth factor C, receptor-binding form; SCDGF; secretory growth factor-like protein; Spinal cord-derived growth factor; VEGF-E
Gene Aliases: FALLOTEIN; PDGFC; SCDGF; UNQ174/PRO200
UniProt ID: (Human) Q9NRA1
Entrez Gene ID: (Human) 56034
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.