Novus Biologicals
Manufacturer Code:NB11060980AF405
Catalog # NBT60980405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0014-5F66 |
Immunogen | This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with human PDGF BB recombinant protein. |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: B chain Becaplermin FLJ12858 PDGF subunit B PDGF2 PDGF-2 platelet-derived growth factor 2 Platelet-derived growth factor beta polypeptide platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog) platelet-derived growth factor subunit B platelet-derived growth factor B chain Platelet-derived growth factor beta polypeptide (oncogene SIS) Proto-oncogene c-Sis SSV; Becaplermin; PDGF subunit B; PDGF, B chain; PDGF-2; platelet-derived growth factor 2; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Proto-oncogene c-Sis
Gene Aliases: c-sis; IBGC5; PDGF-2; PDGF2; PDGFB; SIS; SSV
UniProt ID: (Human) P01127
Entrez Gene ID: (Human) 5155
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.