R&D Systems
Manufacturer Code:MAB2201100
Catalog # MAB2201100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 108104 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PDGF-BB Ser82-Thr190 Accession # P01127 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: B chain Becaplermin C-Sis FLJ12858 IBGC5 PDGF subunit B PDGF2 PDGF-2 PDGFB platelet-derived growth factor 2 Platelet-derived growth factor beta polypeptide platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene ho; Becaplermin; PDGF subunit B; PDGF, B chain; PDGF-2; platelet-derived growth factor 2; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Proto-oncogene c-Sis
Gene Aliases: c-sis; IBGC5; PDGF-2; PDGF2; PDGFB; SIS; SSV
UniProt ID: (Human) P01127
Entrez Gene ID: (Human) 5155
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.