Invitrogen
Manufacturer Code:PA595228
Catalog # PIPA595228
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human PDGF beta recombit protein (Position: S82-T190). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Becaplermin; c-sis; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF-2; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; Platelet-derived growth factor B chain; Platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); Platelet-derived growth factor subunit B; Platelet-derived growth factor subunit B PDGF subunit B PDGF-2 Platelet-derived growth factor B chain Platelet-derived growth factor beta polypeptide Proto-oncogene c-Sis Becaplermin PDGFB PDGF2 SIS; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); Proto-oncogene c-Sis
Gene Aliases: c-sis; IBGC5; PDGF-2; PDGF-B; PDGF2; PDGFB; SIS; SSV
UniProt ID: (Human) P01127, (Mouse) P31240, (Rat) Q05028
Entrez Gene ID: (Human) 5155, (Mouse) 18591, (Rat) 24628
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.