Invitrogen
Manufacturer Code:PA599405
Catalog # PIPA599405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDGFA corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDGF A-chain; PDGF A-chain PDGF subunit A PDGF1 PDGF-1 PDGFA PDGF-A PDGFACP platelet derived growth factor subunit A platelet derived growth factor alpha platelet-derived growth factor A chain platelet-derived growth factor A-chain platelet-derived growth factor alpha chain platelet-derived growth factor alpha polypeptide Platelet-derived growth factor subunit A Rpa1; PDGF subunit A; PDGF-1; platelet derived growth factor, alpha; Platelet-derived growth factor A chain; platelet-derived growth factor A-chain; platelet-derived growth factor alpha chain; Platelet-derived growth factor alpha polypeptide; Platelet-derived growth factor subunit A
Gene Aliases: PDGF-A; PDGF1; PDGFA; PDGFACP; Rpa1
UniProt ID: (Human) P04085, (Rat) P28576, (Mouse) P20033
Entrez Gene ID: (Human) 5154, (Rat) 25266, (Mouse) 18590
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.