Novus Biologicals
Manufacturer Code:NBP273301APCCY7
Catalog # N273301APC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3F3 |
Immunogen | Full length recombinant protein of human PDF (NP_071736) produced in E.coli. |
Conjugate | APC-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.5.1.88 GDF15 MIC1 PDF1A peptide deformylase (mitochondrial) peptide deformylase mitochondrial peptide deformylase-like protein PLAB Polypeptide deformylase; Peptide deformylase, mitochondrial; peptide deformylase-like protein; Polypeptide deformylase
Gene Aliases: PDF; PDF1A
UniProt ID: (Human) Q9HBH1
Entrez Gene ID: (Human) 64146
Molecular Function: hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.