Proteintech
Manufacturer Code:248421AP150UL
Catalog # 50-173-4573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDF Fusion Protein Ag18233 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PDF PDF1A Polypeptide deformylase; Peptide deformylase, mitochondrial; peptide deformylase-like protein; Polypeptide deformylase
Gene Aliases: PDF; PDF1A
UniProt ID: (Human) Q9HBH1
Entrez Gene ID: (Human) 64146
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.