Novus Biologicals
Manufacturer Code:NB3006440.025MG
Catalog # NB018161
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic cyclic peptide common to all PDE8B variants.,This antibody was generated against a common sequence near the C-terminal end that is unique to PDE8B family members (human). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.,Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3',5' cyclic nucleotide phosphodiesterase 8B; 3'-5' cyclic nucleotide phosphodiesterase 8B ADSD Cell proliferation-inducing gene 22 protein EC 3.1.4.17 FLJ11212 high affinity cAMP-specific and IBMX-insensitive 3'-5'-cyclic phosphodiesterase8B hsPDE8B phosphodiesterase 8B PPNAD3; Cell proliferation-inducing gene 22 protein; High affinity cAMP-specific and IBMX-insensitive 3',5'-cyclic phosphodiesterase 8B; HsPDE8B
Gene Aliases: ADSD; PDE8B; PIG22; PPNAD3
UniProt ID: (Human) O95263
Entrez Gene ID: (Human) 8622
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.