Novus Biologicals
Manufacturer Code:NBP29302920UL
Catalog # NB027954
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-87 of human PDE6G (NP_002593.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686C0587 GMP-PDE gamma MGC125749 PDEG phosphodiesterase 6G cGMP-specific rod gamma retinal rod rhodopsin-sensitive cGMP 3&prime-5&prime-cyclic phosphodiesterase subunit gamma rod cG-PDE G RP57; GMP-PDE gamma; phosphodiesterase 6G, cGMP-specific, rod, gamma; Retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit gamma; rod cG-PDE G
Gene Aliases: PDE6G; PDEG; RP57
UniProt ID: (Human) P18545
Entrez Gene ID: (Human) 5148
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.