Novus Biologicals
Manufacturer Code:NBP297479F
Catalog # NB043991
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PDE6D fragment. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GMP-PDE delta; GMP-PDE delta PDED phosphodiesterase 6D cGMP-specific rod delta Protein p17 retinal rod rhodopsin-sensitive cGMP 3'-5'-cyclic phosphodiesterase subunitdelta; phosphodiesterase 6D, cGMP-specific, rod, delta; Protein p17; Retinal rod rhodopsin-sensitive cGMP 3',5'-cyclic phosphodiesterase subunit delta
Gene Aliases: JBTS22; PDE6D; PDED
UniProt ID: (Human) O43924
Entrez Gene ID: (Human) 5147
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.