Invitrogen
Manufacturer Code:PA526732
Catalog # PIPA526732
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 271-300 amino acids from the central region of human PDE6C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGMP phosphodiesterase 6C; COD4 PDEA2 cGMP phosphodiesterase 6C cone cGMP-specific 3&prime5&prime-cyclic phosphodiesterase subunit alpha′ Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'; phosphodiesterase 6C, cGMP-specific, cone, alpha prime
Gene Aliases: ACHM5; COD4; cpfl1; PDE6C; PDEA2
UniProt ID: (Human) P51160, (Mouse) Q91ZQ1
Entrez Gene ID: (Human) 5146, (Mouse) 110855
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.