Novus Biologicals
Manufacturer Code:NBP19850220UL
Catalog # NBP19850220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this antibody is PDE6C - C-terminal region. Peptide sequence LQNNRVEWKSLADEYDAKMKVIEEEAKKQEGGAEKAAEDSGGGDDKKSKT. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGMP phosphodiesterase 6C; cGMP phosphodiesterase 6C COD4 EC 3.1.4 EC 3.1.4.35 PDEA2cone cGMP-specific 3'-5'-cyclic phosphodiesterase subunit alpha' phosphodiesterase 6C cGMP-specific cone alpha prime; Cone cGMP-specific 3',5'-cyclic phosphodiesterase subunit alpha'; phosphodiesterase 6C, cGMP-specific, cone, alpha prime
Gene Aliases: ACHM5; COD4; PDE6C; PDEA2
UniProt ID: (Human) P51160
Entrez Gene ID: (Human) 5146
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.