Proteintech
Manufacturer Code:220631AP150UL
Catalog # 50-173-3709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDE6B Fusion Protein Ag17139 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGMP-phosphodiesterase beta-subunit; CSNB3 GMP PDE beta PDE6B PDEB rd1 RP40; GMP-PDE beta; phosphodiesterase 6B, cGMP-specific, rod, beta; phosphodiesterase, cGMP, rod receptor, beta polypeptide; retinal degeneration 1; rod cGMP-phosphodiesterase beta-subunit; Rod cGMP-specific 3',5'-cyclic phosphodiesterase subunit beta
Gene Aliases: CSNB3; CSNBAD2; Mpb; PDE6B; PDEB; r; rd; rd-1; rd1; rd10; RP40
UniProt ID: (Human) E7ETT3, (Mouse) P23440
Entrez Gene ID: (Human) 5158, (Mouse) 18587
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.