Novus Biologicals
Manufacturer Code:NB3006550.05MG
Catalog # NB300655005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic cyclic peptide (common to all PDE5A variants) (Rat). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: CGB-PDE; CGB-PDE cGMP-binding cGMP-specific 3&prime-5&prime-cyclic nucleotide phosphodiesterase cGMP-binding cGMP-specific phosphodiesterase cGMP-specific 3&prime-5&prime-cyclic phosphodiesterase cGMP-specific phosphodiesterase type 5A CN5A EC 3.1.4 EC 3.1.4.35 PDE5 phosphodiesterase 5A cGMP-specific phosphodiesterase isozyme 5; cGMP-binding cGMP-specific 3',5'-cyclic nucleotide phosphodiesterase; cGMP-binding cGMP-specific phosphodiesterase; cGMP-specific 3',5'-cyclic phosphodiesterase; cGMP-specific phosphodiesterase PDE5A2; cGMP-specific phosphodiesterase type 5A; phosphodiesterase 5A, cGMP-specific; phosphodiesterase isozyme 5
Gene Aliases: CGB-PDE; CN5A; PDE5; PDE5A
UniProt ID: (Human) O76074
Entrez Gene ID: (Human) 8654
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.