Invitrogen
Manufacturer Code:PA521590
Catalog # PIPA521590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 746 and 809 of PDE4D (Uniprot IDNo.Q08499) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP-specific 3',5'-cyclic phosphodiesterase 4D; cAMP-specific phosphodiesterase PDE4D6; DPDE3; DPDE3 HSPDE4D PDE43 PDE4DN2 STRK1 cAMP-specific 3&prime5&prime-cyclic phosphodiesterase 4D cAMP-specific phosphodiesterase PDE4D6 phosphodiesterase 4D cAMP-specific (phosphodiesterase E3 dunce homolog Drosophila); PDE43; phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog, Drosophila); testicular tissue protein Li 136
Gene Aliases: ACRDYS2; DPDE3; HSPDE4D; PDE43; PDE4D; PDE4DN2; STRK1
UniProt ID: (Human) Q08499
Entrez Gene ID: (Human) 5144
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.