Invitrogen
Manufacturer Code:MA525677
Catalog # PIMA525677
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1D12 |
Immunogen | Full length human recombit protein of PDE4B produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cAMP-specific 3',5'-cyclic phosphodiesterase 4B; DPDE4; dunce-like phosphodiesterase E4; PDE32; PDE4B phosphodiesterase 4B cAMP-specific (phosphodiesterase E4 dunce homolog Drosophila) RP5-876C12.1 DKFZp686F2182 DPDE4 MGC126529 PDEIVB OTTHUMP00000046896 cAMP-specific 3&prime5&prime-cyclic phosphodiesterase 4B dunce-like phosphodiesterase E4 phosphodiesterase 4B cAMP-specific phosphodiesterase 4B cAMP-specific (dunce (Drosophila)-homolog phosphodiesterase E4); phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila)
Gene Aliases: DPDE4; dunce; PDE4B; PDEIVB; R74983
UniProt ID: (Human) Q07343
Entrez Gene ID: (Human) 5142, (Mouse) 18578
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.