Invitrogen
Manufacturer Code:PA570201
Catalog # PIPA570201
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of mouse PDE4B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3',5'-cyclic AMP phosphodiesterase; cAMP-specific 3',5'-cyclic phosphodiesterase 4B; DPDE4; dunce-like phosphodiesterase E4; PDE32; PDE4B phosphodiesterase 4B cAMP-specific (phosphodiesterase E4 dunce homolog Drosophila) RP5-876C12.1 DKFZp686F2182 DPDE4 MGC126529 PDEIVB OTTHUMP00000046896 cAMP-specific 3&prime5&prime-cyclic phosphodiesterase 4B dunce-like phosphodiesterase E4 phosphodiesterase 4B cAMP-specific phosphodiesterase 4B cAMP-specific (dunce (Drosophila)-homolog phosphodiesterase E4); Phosphodiesterase 4B cAMP-specific (dunce; phosphodiesterase 4B, cAMP-specific; Phosphodiesterase 4B, cAMP-specific (dunce; phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila)
Gene Aliases: DPDE4; dunce; PDE4/IVb; PDE4B; Pde4B1; Pde4B2; Pde4B3; Pde4B4; PDEIVB; R74983
UniProt ID: (Human) Q07343, (Rat) P14646
Entrez Gene ID: (Dog) 479540, (Human) 5142, (Bovine) 100124505, (Horse) 100053961, (Rabbit) 100355058, (Rat) 24626, (Mouse) 18578, (Guinea pig) 100720296
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.