Novus Biologicals
Manufacturer Code:NBP219745
Catalog # NBP219745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the C-terminus region of human PDE4B. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP-specific (phosphodiesterase E4 dunce homolog cAMP-specific 3'-5'-cyclic phosphodiesterase 4B cAMP-specific phosphodiesterase-4 B isoform DKFZp686F2182 Drosophila) dunce-like phosphodiesterase E4 EC 3.1.4 EC 3.1.4.17 MGC126529 PDE32 PDEIVB phosphodiesterase 4B cAMP-specific phosphodiesterase 4B cAMP-specific (dunce phosphodiesterase 4B cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E4) phosphodiesterase E4 dunce homolog; cAMP-specific 3',5'-cyclic phosphodiesterase 4B; DPDE4; dunce-like phosphodiesterase E4; PDE32; phosphodiesterase 4B, cAMP-specific (phosphodiesterase E4 dunce homolog, Drosophila)
Gene Aliases: DPDE4; PDE4B; PDEIVB
UniProt ID: (Human) Q07343
Entrez Gene ID: (Human) 5142
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.