Novus Biologicals
Manufacturer Code:NBP1433330.1MG
Catalog # NBP14333301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | SMCP3B |
Immunogen | The immunogen for this antibody was PDE3B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CGI-PDE B; CGIP1; CGIP1 CGI-PDE B CGIPDE1 cGMP-inhibited 3'-5'-cyclic phosphodiesterase B Cyclic GMP-inhibited phosphodiesterase B cyclic nucleotide phosphodiesterase EC 3.1.4 EC 3.1.4.17 HcGIP1 phosphodiesterase 3B cGMP-inhibited; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; Cyclic GMP-inhibited phosphodiesterase B; cyclic nucleotide phosphodiesterase; phosphodiesterase 3B, cGMP-inhibited
Gene Aliases: cGIPDE1; HcGIP1; PDE3B
UniProt ID: (Human) Q13370
Entrez Gene ID: (Human) 5140
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.