Invitrogen
Manufacturer Code:14197380
Catalog # 50-129-66
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | SMCP3B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CGI-PDE B; CGI-PDE B CGIP1 CGIPDE1 cGMP-inhibited 3&prime5&prime-cyclic phosphodiesterase B cyclic GMP-inhibited phosphodiesterase B cyclic nucleotide phosphodiesterase phosphodiesterase 3B cGMP inhibited phosphodiesterase 3B cGMP-inhibited; CGIPDE1; cGMP-inhibited 3',5'-cyclic phosphodiesterase B; Cyclic GMP-inhibited phosphodiesterase B
Gene Aliases: 9830102A01Rik; AI847709; Pde3b
UniProt ID: (Mouse) Q61409
Entrez Gene ID: (Mouse) 18576
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.