Novus Biologicals
Manufacturer Code:NB3006730.05MG
Catalog # NB300673005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (selective for only PDE3A variant). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: cAMP phosphodiesterase, myocardial cGMP-inhibited; CGI-PDE A; CGI-PDE CGI-PDE A cGMP-inhibited 3&prime-5&prime-cyclic phosphodiesterase A cyclic GMP inhibited phosphodiesterase A Cyclic GMP-inhibited phosphodiesterase A EC 3.1.4 EC 3.1.4.17 phosphodiesterase 3A cGMP-inhibited; cGMP-inhibited 3',5'-cyclic phosphodiesterase A; Cyclic GMP-inhibited phosphodiesterase A; phosphodiesterase 3A, cGMP-inhibited
Gene Aliases: CGI-PDE; CGI-PDE A; CGI-PDE-A; HTNB; PDE3A
UniProt ID: (Human) Q14432
Entrez Gene ID: (Human) 5139
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.