Abnova
Manufacturer Code:PAB31446
Catalog # 89-998-249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to human PDE3A. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP phosphodiesterase, myocardial cGMP-inhibited; CGI-PDE; CGI-PDE A; cGMP-inhibited 3',5'-cyclic phosphodiesterase A; Cyclic GMP-inhibited phosphodiesterase A; phosphodiesterase 3A, cGMP-inhibited
Gene Aliases: CGI-PDE; CGI-PDE A; CGI-PDE-A; HTNB; PDE3A
UniProt ID: (Human) Q14432
Entrez Gene ID: (Human) 5139
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.