OriGene
Manufacturer Code:TA502320
Catalog # 50-167-3383
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1G9 |
Immunogen | Full length human recombit protein of human PDE2A produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cGMP-dependent 3',5'-cyclic phosphodiesterase; cGMP-stimulated phosphodiesterase 1; cGMP-stimulated phosphodiesterase 2; cGMP-stimulated phosphodiesterase 4; CGS-PDE; CGS-PDE PDE2A1 PED2A4 cGSPDE; Cyclic GMP-stimulated phosphodiesterase; phosphodiesterase 2A, cGMP-stimulated
Gene Aliases: CGS-PDE; cGSPDE; PDE2A; PDE2A1; PED2A4
UniProt ID: (Human) O00408
Entrez Gene ID: (Human) 5138
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.