Invitrogen
Manufacturer Code:PA572298
Catalog # PIPA572298
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 780-810 amino acids of mouse PDE2A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cGMP-dependent 3',5'-cyclic phosphodiesterase; CGS-PDE; CGS-PDE PDE2A1 PED2A4 cGSPDE cGMP-dependent 3&prime5&prime-cyclic phosphodiesterase cGMP-stimulated phosphodiesterase 1 cGMP-stimulated phosphodiesterase 4 cyclic GMP-stimulated phosphodiesterase; Cyclic GMP-stimulated phosphodiesterase
Gene Aliases: CGS-PDE; cGSPDE; Pde2a
UniProt ID: (Mouse) Q922S4
Entrez Gene ID: (Mouse) 207728
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.