Invitrogen
Manufacturer Code:PA5115728
Catalog # PIPA5115728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDE1C corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3',5'-cyclic-AMP phosphodiesterase; 3',5'-cyclic-GMP phosphodiesterase; 3'5'-cyclic-AMP phosphodiesterase 3'5'-cyclic-GMP phosphodiesterase Calcium/calmodulin-dependent 3'5'-cyclic nucleotide phosphodiesterase 1C cam-PDE 1C cyclic nucleotide phosphodiesterase 1 C Hcam3 hCam-3 Human 3'5' cyclic nucleotide phosphodiesterase (HSPDE1C1A) PDE1C phosphodiesterase 1C phosphodiesterase 1C calmodulin-dependent (70kD) phosphodiesterase 1C calmodulin-dependent 70kDa; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1C; Cam-PDE 1C; cyclic nucleotide phosphodiesterase 1 C; Hcam3; Human 3',5' cyclic nucleotide phosphodiesterase (HSPDE1C1A); phosphodiesterase 1C calmodulin-dependent (70kD); phosphodiesterase 1C, calmodulin-dependent 70kDa
Gene Aliases: cam-PDE 1C; hCam-3; Hcam3; PDE1C
UniProt ID: (Human) Q14123, (Mouse) Q64338, (Rat) Q63421
Entrez Gene ID: (Human) 5137, (Mouse) 18575, (Rat) 81742
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.