Novus Biologicals
Manufacturer Code:NBP273278AF700
Catalog # N273278A700
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6E4 |
Immunogen | Full length recombinant protein of human PDE1B (NP_000915) produced in HEK293T cells. |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: 63 kDa Cam-PDE; 63 kDa Cam-PDE calcium/calmodulin-dependent 3&prime-5&prime-cyclic nucleotide phosphodiesterase 1B Cam-PDE 1B EC 3.1.4 EC 3.1.4.17 PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1 phosphodiesterase 1B calmodulin-dependent phosphodiesterase IB calmodulin-dependent Phosphodiesterase-1B presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; calmodulin-stimulated phosphodiesterase PDE1B1; Cam-PDE 1B; epididymis secretory sperm binding protein Li 79p; phosphodiesterase 1B, calmodulin-dependent; presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Gene Aliases: HEL-S-79p; PDE1B; PDE1B1; PDES1B
UniProt ID: (Human) Q01064
Entrez Gene ID: (Human) 5153
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.