Invitrogen
Manufacturer Code:MA517147
Catalog # PIMA517147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5C4A3 |
Immunogen | Purified recombit fragment of human PDE1B (amino acids: 370-536) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 63 kDa Cam-PDE; anti-oxidant protein 2; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; calmodulin-stimulated phosphodiesterase PDE1B1; Cam-PDE 1B; cyclic nucleotide phosphodiesterase (CaM-PDE); epididymis secretory sperm binding protein Li 79p; phosphodiesterase 1B, Ca2+calmodulin dependent; phosphodiesterase 1B, calmodulin-dependent; phosphodiesterase 1B1 Ca2+-calmodulin dependent 63 kDa; phosphodiesterase 1B1, Ca2+-calmodulin dependent, 63 kDa; phosphodiesterase IB calmodulin-dependent; presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Gene Aliases: Aop2; HEL-S-79p; PDE1B; PDE1B1; PDES1B
UniProt ID: (Human) Q01064, (Rat) Q01066
Entrez Gene ID: (Human) 5153, (Rat) 29691
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.