Novus Biologicals
Manufacturer Code:NBP297975B
Catalog # NB050279
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PDE1B fragment. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 63 kDa Cam-PDE; 63 kDa Cam-PDE calcium/calmodulin-dependent 3'-5'-cyclic nucleotide phosphodiesterase 1B Cam-PDE 1B EC 3.1.4 EC 3.1.4.17 PDE1B1calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase PDES1Bcalmodulin-stimulated phosphodiesterase PDE1B1 phosphodiesterase 1B calmodulin-dependent phosphodiesterase IB calmodulin-dependent Phosphodiesterase-1B presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase familyknown as PDE1B; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1B; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; calmodulin-stimulated phosphodiesterase PDE1B1; Cam-PDE 1B; epididymis secretory sperm binding protein Li 79p; phosphodiesterase 1B, calmodulin-dependent; presumed 63kDa form of the type 1 cyclic nucleotide phosphodiesterase family known as PDE1B
Gene Aliases: HEL-S-79p; PDE1B; PDE1B1; PDES1B
UniProt ID: (Human) Q01064
Entrez Gene ID: (Human) 5153
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.