Invitrogen
Manufacturer Code:PA519244
Catalog # PIPA519244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (DLHKNSEDLVNAE) corresponding to the C-terminus amino acids of PDE1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 61 kDa Cam-PDE; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; Cam-PDE 1A; hCam-1; PDE1A phosphodiesterase 1A calmodulin-dependent HCAM1 HSPDE1A MGC26303 3&prime5&prime cyclic nucleotide phosphodiesterase calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase calmodulin-dependent phosphodiesterase phosphodiesterase-1A; phosphodiesterase 1A, calmodulin-dependent
Gene Aliases: CAM-PDE-1A; HCAM-1; HCAM1; HSPDE1A; PDE1A
UniProt ID: (Human) P54750
Entrez Gene ID: (Human) 5136
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.