Invitrogen
Manufacturer Code:PA5103676
Catalog # PIPA5103676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDE1A corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'-RACE clone 8 phosphodiesterase 1A; 3'-RACE clone 8 phosphodiesterase 1A 3-RACE clone 8 phosphodiesterase 1A 3'-RACEclone8phosphodiesterase1A 3-RACEclone8phosphodiesterase1A 61 kDa Cam-PDE AI987702 AW125737 Calcium/calmodulin-dependent 3'5'-cyclic nucleotide phosphodiesterase 1A calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase cam-PDE 1A CAM-PDE-1A HCAM1 HCAM-1 HS MGC26303 HSPDE1A MGC26303 Pde1a phosphodiesterase 1A phosphodiesterase 1A calmodulin-dependent phosphodiesterase-1A; 3-RACE clone 8 phosphodiesterase 1A; 61 kDa Cam-PDE; Calcium/calmodulin-dependent 3',5'-cyclic nucleotide phosphodiesterase 1A; calcium/calmodulin-stimulated cyclic nucleotide phosphodiesterase; Cam-PDE 1A; hCam-1; phosphodiesterase 1A, calmodulin-dependent
Gene Aliases: 3'-RACEclone8phosphodiesterase1A; 3-RACEclone8phosphodiesterase1A; AI987702; AW125737; CAM-PDE-1A; HCAM-1; HCAM1; HSPDE1A; PDE1A
UniProt ID: (Human) P54750, (Mouse) Q61481
Entrez Gene ID: (Human) 5136, (Mouse) 18573, (Rat) 81529
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.