Invitrogen
Manufacturer Code:PA544028
Catalog # PIPA544028
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human PDE12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2',5'-phosphodiesterase 12; 2',5'-phosphodiesterase 12-like protein; 2'-PDE; 2'-phosphodiesterase; 2-PDE; Mitochondrial deadenylase
Gene Aliases: 2'-PDE; 2-PDE; E430028B21Rik; fc58c05; fi42f03; LRRGT00074; PDE12; RGD1310975; wu:fc58c05; wu:fi42f03; zgc:136374
UniProt ID: (Bovine) Q08DF7, (Human) Q6L8Q7, (Mouse) Q3TIU4
Entrez Gene ID: (Horse) 100058532, (Bovine) 538860, (Human) 201626, (Dog) 484714, (Rabbit) 100353603, (Mouse) 211948, (Guinea pig) 100724167, (Rat) 306231, (Zebrafish) 692319
Molecular Function: RNA binding protein exoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.