Invitrogen
Manufacturer Code:PA5116741
Catalog # PIPA5116741
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 17-300 of human PDE12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2',5'-phosphodiesterase 12; 2'-PDE; 2'-phosphodiesterase; 2'5'-phosphodiesterase 12 2'-PDE 2-PDE 2'-phosphodiesterase E430028B21Rik LRRGT00074 Mitochondrial deadenylase PDE12 phosphodiesterase 12 RGD1310975; 2-PDE; Mitochondrial deadenylase
Gene Aliases: 2'-PDE; 2-PDE; E430028B21Rik; LRRGT00074; PDE12; RGD1310975
UniProt ID: (Human) Q6L8Q7, (Mouse) Q3TIU4
Entrez Gene ID: (Human) 201626, (Mouse) 211948, (Rat) 306231
Molecular Function:
RNA binding protein
exoribonuclease
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.