Invitrogen
Manufacturer Code:PA518767
Catalog # PIPA518767
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NSFKESMEKSSYSD) corresponding to the internal amino acids of PDE11A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP and cGMP cyclic nucleotide phosphodiesterase 11A; cAMP and cGMP phosphodiesterase 11A; Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A; PDE11A phosphodiesterase 11A FLJ23693 MGC133355 MGC133356 PPNAD2 cAMP and cGMP cyclic nucleotide phosphodiesterase 11A cyclic nucleotide phosphodiesterase 11A dual 3&prime5&prime-cyclic-AMP and -GMP phosphodiesterase 11A
Gene Aliases: PDE11A; PPNAD2
UniProt ID: (Human) Q9HCR9
Entrez Gene ID: (Human) 50940
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.