Invitrogen
Manufacturer Code:PA5115727
Catalog # PIPA5115727
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PDE11A corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 6330414F14Rik A630086N24Rik cAMP and cGMP cyclic nucleotide phosphodiesterase 11A cAMP and cGMP phosphodiesterase 11A Dual 3'5'-cyclic-AMP and -GMP phosphodiesterase 11A FLJ23693 Gm350 MGC133355 MGC133356 PDE11A PDE11A1 phosphodiesterase 11A PPNAD2; cAMP and cGMP cyclic nucleotide phosphodiesterase 11A; cAMP and cGMP phosphodiesterase 11A; Dual 3',5'-cyclic-AMP and -GMP phosphodiesterase 11A
Gene Aliases: 6330414F14Rik; A630086N24Rik; Gm350; PDE11A; PDE11A1; PPNAD2
UniProt ID: (Human) Q9HCR9, (Mouse) P0C1Q2, (Rat) Q8VID6
Entrez Gene ID: (Human) 50940, (Mouse) 241489, (Rat) 140928
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.