Novus Biologicals
Manufacturer Code:NBP273273JF549
Catalog # N273273J549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C9 |
Immunogen | Full-length protein expressed in 293T cell transfected with human PDE10A expression vector. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cAMP and cAMP-inhibited cGMP 3&prime-5&prime-cyclic phosphodiesterase 10A dJ416F21.1 (phosphodiesterase 10A) EC 3.1.4 EC 3.1.4.17 EC 3.1.4.35 FLJ11894 FLJ25677 HSPDE10A phosphodiesterase 10A phosphodiesterase 10A1 (PDE10A1); cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A; dJ416F21.1 (phosphodiesterase 10A); phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A7 (PDE10A7)
Gene Aliases: ADSD2; HSPDE10A; IOLOD; LINC00473; PDE10A; PDE10A19
UniProt ID: (Human) Q9Y233
Entrez Gene ID: (Human) 10846
Molecular Function: hydrolase phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.