Invitrogen
Manufacturer Code:PA531293
Catalog # PIPA531293
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 547 and 779 of Human PDE10A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cAMP and cAMP-inhibited cGMP 3',5'-cyclic phosphodiesterase 10A; dJ416F21.1 (phosphodiesterase 10A); HSPDE10A cAMP and cAMP-inhibited cGMP 3&prime5&prime-cyclic phosphodiesterase 10A dJ416F21.1 (phosphodiesterase 10A) phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A1 (PDE10A1); phosphodiesterase 10A7 (PDE10A7)
Gene Aliases: ADSD2; HSPDE10A; IOLOD; LINC00473; PDE10A; PDE10A19
UniProt ID: (Human) Q9Y233
Entrez Gene ID: (Human) 10846
Molecular Function:
hydrolase
phosphodiesterase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.