OriGene
Manufacturer Code:TA809714
Catalog # 50-168-2981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI6G6 |
Immunogen | Full length human recombit protein of human PDCL3 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HTPHLP; HTPHLP PHLP2A PHLP3 VIAF VIAF1; IAP-associated factor VIAF1; phosducin-like 3; Phosducin-like protein 3; PhPL3; VIAF-1; Viral IAP-associated factor 1
Gene Aliases: HTPHLP; PDCL3; PHLP2A; PHLP3; VIAF; VIAF1
UniProt ID: (Human) Q9H2J4
Entrez Gene ID: (Human) 79031
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.