Invitrogen
Manufacturer Code:PA553810
Catalog # PIPA553810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PDCL3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HTPHLP; IAP-associated factor VIAF1; phosducin-like 3; Phosducin-like protein 3; PhPL3; VIAF PHLP3 VIAF1 HTPHLP PHLP2A PhLP2A Phosducin-like protein 3 VIAF-1; VIAF-1; Viral IAP-associated factor 1
Gene Aliases: HTPHLP; PDCL3; PHLP2A; PHLP3; VIAF; VIAF1
UniProt ID: (Human) Q9H2J4
Entrez Gene ID: (Human) 79031
Molecular Function: G-protein modulator chaperone enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.