Novus Biologicals
Manufacturer Code:NBP180627
Catalog # NBP180627
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:IGPLVFGGMNLTRDELEWKLSESGAIMTDLEENPKKPIEDVLLSSVRRSVLMKRDSDSEGD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HTPHLP; HTPHLP IAP-associated factor VIAF1 MGC3062 PHLP3 phosducin-like 3 phosducin-like protein 3 PhPL3 VIAF VIAF-1 VIAF1PHLP2A Viral IAP-associated factor 1; IAP-associated factor VIAF1; phosducin-like 3; Phosducin-like protein 3; PhPL3; VIAF-1; Viral IAP-associated factor 1
Gene Aliases: HTPHLP; PDCL3; PHLP2A; PHLP3; VIAF; VIAF1
UniProt ID: (Human) Q9H2J4
Entrez Gene ID: (Human) 79031
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.