Invitrogen
Manufacturer Code:PA538056
Catalog # PIPA538056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 18 amino acid peptide near the carboxy terminus of human PDCL2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GCPHLP Phosducin-like protein 2; MgcPhLP; phosducin-like 2; Phosducin-like protein 2; Phosducin-like protein similar 1; protein B
Gene Aliases: 1700010B22Rik; 1700016K07Rik; GCPHLP; Mgcphlp; PDCL2; RGD1559712; Tphlp
UniProt ID: (Human) Q8N4E4, (Mouse) Q78Y63
Entrez Gene ID: (Human) 132954, (Rat) 498352, (Mouse) 79455
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.