Invitrogen
Manufacturer Code:PA5113181
Catalog # PIPA5113181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Phosducin-like protein 2 protein (1-241AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1700010B22Rik 1700016K07Rik GCPHLP MgcPhLP Pdcl2 phosducin like 2 phosducin-like 2 Phosducin-like protein 2 Phosducin-like protein similar 1 protein B RGD1559712 Tphlp; phosducin-like 2; Phosducin-like protein 2
Gene Aliases: GCPHLP; PDCL2; RGD1559712
UniProt ID: (Human) Q8N4E4
Entrez Gene ID: (Human) 132954, (Rat) 498352
Molecular Function:
G-protein modulator
chaperone
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.