Invitrogen
Manufacturer Code:MA536229
Catalog # PIMA536229
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 14D10 |
Immunogen | E.coli-derived human PDCD6IP recombinant protein (Position: A2-D330). Human PDCD6IP shares96.7% and 95.2% aminoacid (aa)sequenceidentitywith mouseand rat PDCD6IP respectively. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI480591 aip1 ALG-2 interacting protein 1 ALG-2 interacting protein X ALG-2-interacting protein 1 Alg2-interacting protein 1 ALG-2-interacting protein X Alg2-interacting protein X alix apoptosis-linked gene 2-interacting protein X AW544830 C76364 dopamine receptor interacting protein 4 DRIP4 E2F1-inducible protein Eig2 HP95 KIAA1375 MGC17003 mKIAA1375 PDCD6-interacting protein PDCD6IP pdcd6ip.L programmed cell death 6 interacting protein programmed cell death 6 interacting protein L homeolog programmed cell death 6-interacting protein putative signal tranduction protein Xp95 RGD1561176 Signal transduction protein Xp95 XELAEV_18031317mg xp95; ALG-2 interacting protein 1; ALG-2 interacting protein X; ALG-2-interacting protein 1; ALG-2-interacting protein X; Alg2-interacting protein 1; Alg2-interacting protein X; apoptosis-linked gene 2-interacting protein X; dopamine receptor interacting protein 4; E2F1-inducible protein; Eig2; Hp95; PDCD6-interacting protein; Programmed cell death 6-interacting protein
Gene Aliases: AI480591; AIP1; ALIX; AW544830; C76364; DRIP4; Eig2; HP95; KIAA1375; mKIAA1375; PDCD6IP; RGD1561176
UniProt ID: (Human) Q8WUM4, (Rat) Q9QZA2, (Mouse) Q9WU78
Entrez Gene ID: (Human) 10015, (Rat) 501083, (Mouse) 18571
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.