Proteintech
Manufacturer Code:124221AP150UL
Catalog # 50-172-7971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDCD6IP Fusion Protein Ag3074 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIP1 ALG 2 interacting protein 1 Alix DRIP4 HP95 KIAA1375 PDCD6 interacting protein PDCD6IP; ALG-2 interacting protein 1; ALG-2 interacting protein X; ALG-2-interacting protein 1; ALG-2-interacting protein X; Alg2-interacting protein 1; Alg2-interacting protein X; apoptosis-linked gene 2-interacting protein X; dopamine receptor interacting protein 4; E2F1-inducible protein; Eig2; Hp95; PDCD6-interacting protein; Programmed cell death 6-interacting protein
Gene Aliases: AI480591; AIP1; ALIX; AW544830; C76364; DRIP4; Eig2; HP95; KIAA1375; mKIAA1375; PDCD6IP; RGD1561176
UniProt ID: (Human) Q8WUM4, (Rat) Q9QZA2, (Mouse) Q9WU78
Entrez Gene ID: (Human) 10015, (Rat) 501083, (Mouse) 18571
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.