Novus Biologicals
Manufacturer Code:NBP267024
Catalog # NBP267024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JA80-83 |
Immunogen | Full length protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: H731nuclear antigen H731 MGC33046 MGC33047 Neoplastic transformation inhibitor protein Nuclear antigen H731-like programmed cell death 4 (neoplastic transformation inhibitor) programmed cell death protein 4 Protein 197/15a; Neoplastic transformation inhibitor protein; nuclear antigen H731; Nuclear antigen H731-like; Programmed cell death protein 4; Protein 197/15a
Gene Aliases: H731; PDCD4
UniProt ID: (Human) Q53EL6
Entrez Gene ID: (Human) 27250
Molecular Function:
RNA binding protein
nucleic acid binding
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.