R&D Systems
Manufacturer Code:MAB7019
Catalog # MAB7019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 808508 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PDCD4 Lys212-His358 Accession # Q53EL6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: H731 H731nuclear antigen H731 MGC33046 MGC33047 Neoplastic transformation inhibitor protein Nuclear antigen H731-like programmed cell death 4 (neoplastic transformation inhibitor) programmed cell death protein 4 Protein 197/15a; Neoplastic transformation inhibitor protein; nuclear antigen H731; Nuclear antigen H731-like; Programmed cell death protein 4; Protein 197/15a
Gene Aliases: H731; PDCD4
UniProt ID: (Human) Q53EL6
Entrez Gene ID: (Human) 27250
Molecular Function: RNA binding protein nucleic acid binding translation elongation factor translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.