Proteintech
Manufacturer Code:125871AP150UL
Catalog # 50-172-8088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PDCD4 Fusion Protein Ag3288 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Death up-regulated gene protein; death-upregulated; H731 Nuclear antigen H731 like PDCD4 Protein 197/15a; Neoplastic transformation inhibitor protein; nuclear antigen H731; Nuclear antigen H731-like; Programmed cell death protein 4; Protein 197/15a; Protein MA-3; Topoisomerase-inhibitor suppressed protein
Gene Aliases: D19Ucla1; Dug; H731; Ma3; PDCD4; Tis
UniProt ID: (Human) Q53EL6, (Rat) Q9JID1, (Mouse) Q61823
Entrez Gene ID: (Human) 27250, (Rat) 64031, (Mouse) 18569
Molecular Function:
RNA binding protein
nucleic acid binding
translation elongation factor
translation factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.