Invitrogen
Manufacturer Code:PA555388
Catalog # PIPA555388
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PDCD2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Programmed cell death protein 2; RP8 ZMYND7 programmed cell death protein 2 zinc finger MYND domain-containing protein 7 zinc finger protein Rp-8; Zinc finger MYND domain-containing protein 7; Zinc finger protein Rp-8
Gene Aliases: AI528543; Pcd2; PDCD2; RP-8; RP8; ZMYND7
UniProt ID: (Human) Q16342, (Rat) P47816, (Mouse) P46718
Entrez Gene ID: (Human) 5134, (Rat) 58934, (Mouse) 18567
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.