Invitrogen
Manufacturer Code:PA579791
Catalog # PIPA579791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human PDCD10 recombit protein (Position: M1-A212). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoptosis-related protein 15; Cerebral cavernous malformations 3 protein; Programmed cell death protein 10; Programmed cell death protein 10 Cerebral cavernous malformations 3 protein TF-1 cell apoptosis-related protein 15 PDCD10 CCM3 TFAR15; TF-1 cell apoptosis-related protein 15
Gene Aliases: 2410003B13Rik; CCM3; PDCD10; Tfa15; TFAR15
UniProt ID: (Human) Q9BUL8, (Rat) Q6NX65, (Mouse) Q8VE70
Entrez Gene ID: (Human) 11235, (Rat) 494345, (Mouse) 56426
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.