Invitrogen
Manufacturer Code:PA576681
Catalog # PIPA576681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PDCD10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Apoptosis related protein 15CCM3Cerebral cavernous malformations 3 proteinMGC1212MGC24477PDC10&lowbarHUMANPDCD 10PDCD10Programmed cell death 10Programmed cell death protein 10TF 1 cell apoptosis related protein 15TF-1 cell apoptosis-related protein 15TFAR15; apoptosis-related protein 15; Cerebral cavernous malformations 3 protein; Programmed cell death protein 10; TF-1 cell apoptosis-related protein 15
Gene Aliases: 2410003B13Rik; CCM3; PDCD10; Tfa15; TFAR15
UniProt ID: (Human) Q9BUL8, (Mouse) Q8VE70
Entrez Gene ID: (Human) 11235, (Mouse) 56426
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.