Novus Biologicals
Manufacturer Code:NBP297685DL594
Catalog # NB046538
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human PDCD10 fragment. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: apoptosis-related protein 15; apoptosis-related protein 15 cerebral cavernous malformations 3 MGC1212 MGC24477 programmed cell death 10 programmed cell death protein 10 TF-1 cell apoptosis-related protein 15 TFAR15CCM3Cerebral cavernous malformations 3 protein; Cerebral cavernous malformations 3 protein; Programmed cell death protein 10; TF-1 cell apoptosis-related protein 15
Gene Aliases: CCM3; PDCD10; TFAR15
UniProt ID: (Human) Q9BUL8
Entrez Gene ID: (Human) 11235
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.